Assay Details
Gene Symbol: Mx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134350.2 | NP_599177.2 | 5 - 6 | 705 | 132 | |
XM_006248150.2 | XP_006248212.1 | 7 - 8 | 849 | 132 | ||
XM_008768568.1 | XP_008766790.1 | 7 - 8 | 783 | 132 | ||
GenBank mRNA | BC099808.1 | - | 5 - 6 | 705 | 132 | |
X52712.1 | - | 5 - 6 | 780 | 132 | ||
X52713.1 | - | 4 - 5 | 585 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
