Assay Details
Gene Symbol: Pnlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013161.2 | NP_037293.1 | 7 - 8 | 720 | 97 | |
GenBank mRNA | D88534.1 | - | 6 - 7 | 692 | 97 | |
D88535.1 | - | 6 - 7 | 692 | 97 | ||
M58369.1 | - | 7 - 8 | 721 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
