Assay Details
Gene Symbol: Fam204a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107448.2 | NP_001100918.1 | 2 - 3 | 400 | 128 | |
XM_006231673.2 | XP_006231735.1 | 2 - 3 | 1216 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
