Assay Details
Gene Symbol: Cyp17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012753.2 | NP_036885.1 | 4 - 5 | 775 | 85 | |
XM_006231434.2 | XP_006231496.1 | 5 - 6 | 1098 | 85 | ||
XM_006231435.2 | XP_006231497.1 | 5 - 6 | 1067 | 85 | ||
GenBank mRNA | BC078898.1 | - | 4 - 5 | 775 | 85 | |
M22204.1 | - | 4 - 5 | 777 | 85 | ||
M31681.1 | - | 4 - 5 | 787 | 85 | ||
X14086.1 | - | 4 - 5 | 777 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
