Assay Details
Gene Symbol: Htr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024394.2 | NP_077370.2 | 3 - 4 | 392 | 86 | |
XM_006243021.2 | XP_006243083.1 | 3 - 4 | 406 | 86 | ||
GenBank mRNA | CB583270.1 | - | 3 - 4 | 392 | 86 | |
D49395.1 | - | 3 - 4 | 283 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
