Assay Details
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017075.2 | NP_058771.2 | 2 - 3 | 189 | 97 | |
GenBank mRNA | D00512.1 | - | 2 - 3 | 121 | 97 | |
D13921.1 | - | 2 - 3 | 121 | 97 | ||
FM065829.1 | - | 2 - 3 | 189 | 97 | ||
FQ217320.1 | - | 2 - 3 | 127 | 97 | ||
FQ228108.1 | - | 2 - 3 | 176 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
