Assay Details
Gene Symbol: Dbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109437.2 | NP_001102907.1 | 4 - 5 | 597 | 188 | |
GenBank mRNA | BC166797.1 | - | 4 - 5 | 597 | 188 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
