Assay Details
Gene Symbol: Slc25a38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030032.1 | NP_001025203.1 | 6 - 7 | 948 | 64 | |
XM_006244089.2 | XP_006244151.1 | 6 - 7 | 2849 | 64 | ||
GenBank mRNA | BC099762.1 | - | 6 - 7 | 948 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
