Assay Details
Gene Symbol: Ctsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012939.1 | NP_037071.1 | 2 - 3 | 126 | 82 | |
XM_008766440.1 | XP_008764662.1 | 2 - 3 | 177 | 82 | ||
GenBank mRNA | BC085352.1 | - | 2 - 3 | 141 | 82 | |
FQ210730.1 | - | 2 - 3 | 136 | 82 | ||
FQ213183.1 | - | 2 - 3 | 136 | 82 | ||
FQ219177.1 | - | 2 - 3 | 136 | 82 | ||
FQ219281.1 | - | 2 - 3 | 136 | 82 | ||
FQ219285.1 | - | 2 - 3 | 136 | 82 | ||
FQ219548.1 | - | 2 - 3 | 136 | 82 | ||
FQ219611.1 | - | 2 - 3 | 154 | 82 | ||
FQ223707.1 | - | 2 - 3 | 132 | 82 | ||
FQ228686.1 | - | 2 - 3 | 196 | 82 | ||
FQ228906.1 | - | 2 - 3 | 143 | 82 | ||
FQ228938.1 | - | 2 - 3 | 133 | 82 | ||
FQ228956.1 | - | 2 - 3 | 163 | 82 | ||
M38135.1 | - | 2 - 3 | 121 | 82 | ||
Y00708.1 | - | 2 - 3 | 126 | 82 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the M38135.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
