Assay Details
Gene Symbol: Ctsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012939.1 | NP_037071.1 | 8 - 9 | 633 | 65 | |
XM_008766440.1 | XP_008764662.1 | 8 - 9 | 684 | 65 | ||
GenBank mRNA | BC085352.1 | - | 8 - 9 | 648 | 65 | |
FQ210730.1 | - | 8 - 9 | 643 | 65 | ||
FQ213183.1 | - | 8 - 9 | 643 | 65 | ||
FQ219177.1 | - | 8 - 9 | 643 | 65 | ||
FQ219285.1 | - | 8 - 9 | 643 | 65 | ||
FQ219548.1 | - | 8 - 9 | 643 | 65 | ||
FQ219611.1 | - | 8 - 9 | 661 | 65 | ||
FQ220588.1 | - | 5 - 6 | 412 | 65 | ||
FQ223707.1 | - | 8 - 9 | 639 | 65 | ||
FQ228686.1 | - | 8 - 9 | 703 | 65 | ||
FQ228906.1 | - | 8 - 9 | 650 | 65 | ||
FQ228938.1 | - | 8 - 9 | 640 | 65 | ||
FQ228956.1 | - | 8 - 9 | 670 | 65 | ||
M38135.1 | - | 8 - 9 | 627 | 65 | ||
Y00708.1 | - | 8 - 9 | 633 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
