Assay Details
Gene Symbol: Ctsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012939.1 | NP_037071.1 | 9 - 10 | 694 | 65 | |
XM_008766440.1 | XP_008764662.1 | 9 - 10 | 745 | 65 | ||
GenBank mRNA | BC085352.1 | - | 9 - 10 | 709 | 65 | |
FQ210730.1 | - | 9 - 10 | 704 | 65 | ||
FQ213183.1 | - | 9 - 10 | 704 | 65 | ||
FQ219177.1 | - | 9 - 10 | 704 | 65 | ||
FQ219281.1 | - | 9 - 10 | 704 | 65 | ||
FQ219285.1 | - | 9 - 10 | 704 | 65 | ||
FQ219548.1 | - | 9 - 10 | 704 | 65 | ||
FQ220588.1 | - | 6 - 7 | 473 | 65 | ||
FQ223707.1 | - | 9 - 10 | 700 | 65 | ||
FQ228686.1 | - | 9 - 10 | 764 | 65 | ||
FQ228938.1 | - | 9 - 10 | 701 | 65 | ||
FQ228956.1 | - | 9 - 10 | 731 | 65 | ||
M38135.1 | - | 9 - 10 | 688 | 65 | ||
Y00708.1 | - | 9 - 10 | 694 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
