Assay Details
Gene Symbol: Rassf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007754.1 | NP_001007755.1 | 2 - 3 | 287 | 65 | |
NM_001037555.1 | NP_001032644.1 | 3 - 4 | 479 | 65 | ||
GenBank mRNA | AB202124.1 | - | 3 - 4 | 479 | 65 | |
BC079237.1 | - | 2 - 3 | 287 | 65 | ||
FQ233012.1 | - | 2 - 3 | 327 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
