Assay Details
Gene Symbol: Hemk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106853.1 | NP_001100323.1 | 6 - 7 | 797 | 88 | |
XM_006243728.2 | XP_006243790.1 | 7 - 8 | 1087 | 88 | ||
XM_006243729.2 | XP_006243791.1 | 7 - 8 | 1407 | 88 | ||
XM_006243730.2 | XP_006243792.1 | 6 - 7 | 1162 | 88 | ||
XM_008766502.1 | XP_008764724.1 | 7 - 8 | 1022 | 88 | ||
GenBank mRNA | BC161944.1 | - | 6 - 7 | 791 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
