Assay Details
Gene Symbol: Cyb5r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133427.3 | NP_596918.3 | 4 - 5 | 553 | 64 | |
XM_008766437.1 | XP_008764659.1 | 4 - 5 | 712 | 64 | ||
GenBank mRNA | AF307840.1 | - | 3 - 4 | 312 | 64 | |
AY321370.1 | - | 3 - 4 | 312 | 64 | ||
AY321371.1 | - | 3 - 4 | 312 | 64 | ||
BC080240.1 | - | 4 - 5 | 531 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
