Assay Details
Gene Symbol: Elp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108782.2 | NP_001102252.1 | 3 - 4 | 230 | 69 | |
GenBank mRNA | BC166773.1 | - | 3 - 4 | 230 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
