Assay Details
Gene Symbol: Ppp2r3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012202.2 | NP_001012202.2 | 8 - 9 | 3138 | 106 | |
XM_006243633.2 | XP_006243695.1 | 9 - 10 | 3440 | 106 | ||
XM_006243634.2 | XP_006243696.1 | 8 - 9 | 1864 | 106 | ||
GenBank mRNA | BC087097.1 | - | 8 - 9 | 1213 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
