Assay Details
Gene Symbol: Mrpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106852.1 | NP_001100322.1 | 2 - 3 | 461 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
