Assay Details
Gene Symbol: Slc17a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009713.2 | NP_001009713.1 | 2 - 3 | 331 | 81 | |
GenBank mRNA | AY800277.1 | - | 2 - 3 | 330 | 81 | |
BC097482.1 | - | 2 - 3 | 331 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
