Assay Details
Gene Symbol: Gsta4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106840.1 | NP_001100310.1 | 341 | 87 | ||
GenBank mRNA | BC157819.1 | - | 311 | 87 | ||
FJ179399.1 | - | 3 - 4 | 274 | 87 | ||
FQ230022.1 | - | 321 | 87 | |||
FQ233696.1 | - | 323 | 87 | |||
FQ233840.1 | - | 323 | 87 | |||
X62660.1 | - | 302 | 87 |
Gene Symbol: LOC100909835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750542.3 | XP_003750590.1 | 1 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
