Assay Details
Gene Symbol: Gsta4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106840.1 | NP_001100310.1 | 145 | 145 | ||
GenBank mRNA | BC157819.1 | - | 115 | 145 | ||
FJ179399.1 | - | 1 - 2 | 78 | 145 | ||
FQ230022.1 | - | 125 | 145 | |||
FQ233696.1 | - | 127 | 145 | |||
FQ233840.1 | - | 127 | 145 | |||
X62660.1 | - | 106 | 145 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
