Assay Details
Gene Symbol: Myrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182844.1 | NP_878264.1 | 12 - 13 | 2253 | 99 | |
XM_006244110.2 | XP_006244172.1 | 12 - 13 | 2293 | 99 | ||
XM_008766684.1 | XP_008764906.1 | 13 - 14 | 2371 | 99 | ||
XM_008766685.1 | XP_008764907.1 | 14 - 15 | 2485 | 99 | ||
XM_008766686.1 | XP_008764908.1 | 12 - 13 | 2295 | 99 | ||
XM_008766687.1 | XP_008764909.1 | 11 - 12 | 2766 | 99 | ||
GenBank mRNA | AY331984.1 | - | 12 - 13 | 2253 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
