Assay Details
Gene Symbol: Atrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106859.1 | NP_001100329.1 | 10 - 11 | 2064 | 62 | |
XM_006243764.2 | XP_006243826.1 | 10 - 11 | 2063 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
