Assay Details
Gene Symbol: Nprl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025744.2 | NP_001020915.2 | 2 - 3 | 308 | 61 | |
GenBank mRNA | BC098932.1 | - | 2 - 3 | 287 | 61 | |
CV077924.1 | - | 2 - 3 | 308 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
