Assay Details
Gene Symbol: Nprl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025744.2 | NP_001020915.2 | 4 - 5 | 587 | 61 | |
GenBank mRNA | CV077924.1 | - | 4 - 5 | 587 | 61 | |
FQ209669.1 | - | 3 - 4 | 661 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
