Assay Details
Gene Symbol: Nprl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025744.2 | NP_001020915.2 | 8 - 9 | 950 | 65 | |
GenBank mRNA | BC098932.1 | - | 5 - 6 | 1412 | 65 | |
BI285715.1 | - | 3 - 4 | 65 | |||
DV714395.1 | - | 4 - 5 | 358 | 65 | ||
FQ209669.1 | - | 7 - 8 | 1023 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
