Assay Details
Gene Symbol: Cdhr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243871.2 | XP_006243933.1 | 13 - 14 | 1980 | 69 | |
XM_006243872.2 | XP_006243934.1 | 13 - 14 | 1979 | 69 | ||
XM_006243880.2 | XP_006243942.1 | 9 - 10 | 1591 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
