Assay Details
Gene Symbol: Cdhr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243871.2 | XP_006243933.1 | 9 - 10 | 1390 | 60 | |
XM_006243872.2 | XP_006243934.1 | 9 - 10 | 1389 | 60 | ||
XM_006243880.2 | XP_006243942.1 | 5 - 6 | 1001 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
