Assay Details
Gene Symbol: Ip6k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021660.2 | NP_067692.2 | 5 - 6 | 861 | 70 | |
XM_008766564.1 | XP_008764786.1 | 4 - 5 | 827 | 70 | ||
XM_008766565.1 | XP_008764787.1 | 3 - 4 | 537 | 70 | ||
XM_008766566.1 | XP_008764788.1 | 5 - 6 | 896 | 70 | ||
GenBank mRNA | BC083574.1 | - | 5 - 6 | 861 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
