Assay Details
Gene Symbol: Bckdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019267.1 | NP_062140.1 | 1 - 2 | 219 | 91 | |
GenBank mRNA | BC158861.1 | - | 1 - 2 | 212 | 91 | |
CK473898.1 | - | 1 - 2 | 219 | 91 | ||
FQ213896.1 | - | 1 - 2 | 201 | 91 | ||
FQ226240.1 | - | 1 - 2 | 202 | 91 | ||
M94040.1 | - | 1 - 2 | 130 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
