Assay Details
Gene Symbol: Bckdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019267.1 | NP_062140.1 | 6 - 7 | 764 | 132 | |
XM_006243450.2 | XP_006243512.1 | 7 - 8 | 821 | 132 | ||
GenBank mRNA | BC158861.1 | - | 6 - 7 | 757 | 132 | |
CB547976.1 | - | 2 - 3 | 185 | 132 | ||
FQ213896.1 | - | 6 - 7 | 746 | 132 | ||
M94040.1 | - | 6 - 7 | 675 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
