Assay Details
Gene Symbol: Atp1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012913.1 | NP_037045.1 | 6 - 7 | 818 | 171 | |
GenBank mRNA | BC061719.1 | - | 6 - 7 | 821 | 171 | |
D84450.1 | - | 6 - 7 | 818 | 171 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
potassium ion transport
sodium ion transport
cellular sodium ion homeostasis
metal ion transport
cellular potassium ion homeostasis
positive regulation of ATPase activity
sodium ion export across plasma membrane
protein stabilization
protein localization to plasma membrane
membrane repolarization
positive regulation of potassium ion transmembrane transporter activity
positive regulation of sodium ion export across plasma membrane
positive regulation of potassium ion import across plasma membrane
potassium ion import across plasma membrane
ATPase activator activity
P-type sodium:potassium-exchanging transporter activity
ATPase binding
potassium ion transport
sodium ion transport
cellular sodium ion homeostasis
metal ion transport
cellular potassium ion homeostasis
positive regulation of ATPase activity
sodium ion export across plasma membrane
protein stabilization
protein localization to plasma membrane
membrane repolarization
positive regulation of potassium ion transmembrane transporter activity
positive regulation of sodium ion export across plasma membrane
positive regulation of potassium ion import across plasma membrane
potassium ion import across plasma membrane
ATPase activator activity
P-type sodium:potassium-exchanging transporter activity
ATPase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P555125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45208 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P555125 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P555125 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P555125 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P555125 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700934989 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57353 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12883 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102732 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700934989 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10919379 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10910192 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919380 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919378 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919381 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10919377 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10910191 | - | - |
Rat Genome 230 2.0 Array | 1398300_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398300_at | - | - |
Affymetrix HT Rat Focus Array | 1398300_at | - | - |
Affymetrix HT Rat Focus Array | 1398300_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800025290.rn.1 | TC0800003168.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800025293.rn.1 | TC0800003168.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800025288.rn.1 | TC0800003168.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800025291.rn.1 | TC0800003168.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800025289.rn.1 | TC0800003168.rn.1 | RTA TC |
Rat Expression Set 230 | 1398300_PM_at | - | - |
Rat Expression Set 230 | 1398300_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919377 | - | - |
Rat Genome U34 Set | D84450_at | - | - |
Rat Genome U34 Set | rc_AA943304_g_at | - | - |
Rat Genome U34 Set | rc_AA943304_at | - | - |
Rat Genome U34 Set | rc_AA943304_g_at | - | - |
Rat Genome U34 Set | D84450_at | - | - |
Rat Genome U34 Set | D84450_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21044593 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369650 | - | - |
RatRef-12 Expression BeadChip | ILMN_69740 | - | - |
