Assay Details
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013252.1 | NP_001013270.1 | 3 - 4 | 339 | 131 | |
GenBank mRNA | AY325214.1 | - | 2487 | 131 | ||
AY325230.1 | - | 2490 | 131 | |||
AY325241.1 | - | 2487 | 131 | |||
BC098951.1 | - | 331 | 131 | |||
BF525252.1 | - | 345 | 131 | |||
BQ204036.1 | - | 4 - 5 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
