Assay Details
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013252.1 | NP_001013270.1 | 4 - 5 | 424 | 68 | |
GenBank mRNA | AY325214.1 | - | 2572 | 68 | ||
AY325230.1 | - | 2575 | 68 | |||
AY325241.1 | - | 2572 | 68 | |||
BC098951.1 | - | 416 | 68 | |||
BF523222.1 | - | 1 - 2 | 90 | 68 | ||
BQ204036.1 | - | 3 - 4 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
