Assay Details
Gene Symbol: Mlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766417.1 | XP_008764639.1 | 10 - 11 | 2815 | 147 | |
XM_008766418.1 | XP_008764640.1 | 9 - 10 | 2659 | 147 | ||
XM_008766420.1 | XP_008764642.1 | 10 - 11 | 2623 | 147 | ||
GenBank mRNA | BC092625.1 | - | 2235 | 147 | ||
FQ224108.1 | - | 2478 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
