Assay Details
Gene Symbol: Cwc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024987.1 | NP_001020158.1 | 6 - 7 | 691 | 116 | |
XM_006242520.2 | XP_006242582.1 | 6 - 7 | 722 | 116 | ||
GenBank mRNA | BC091396.1 | - | 6 - 7 | 691 | 116 | |
FQ224752.1 | - | 6 - 7 | 661 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
