Assay Details
Gene Symbol: Chordc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108128.1 | NP_001101598.1 | 10 - 11 | 996 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P941132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163568 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P941132 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P941132 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P941132 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P152329 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P941132 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P754987 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24655 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10908142 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10908134 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10908135 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10908143 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10908137 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10908141 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10908144 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10908138 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10908139 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10908136 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10908140 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10908133 | - | - |
Rat Genome 230 2.0 Array | 1383160_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383160_at | - | - |
Affymetrix HT Rat Focus Array | 1383160_at | - | - |
Affymetrix HT Rat Focus Array | 1383160_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800000848.rn.1 | TC0800000140.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800000850.rn.1 | TC0800000140.rn.1 | RTA TC |
Rat Expression Set 230 | 1383160_PM_at | - | - |
Rat Expression Set 230 | 1383160_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10908133 | - | - |
Rat Genome U34 Set | rc_AA892238_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21491251 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367875 | - | - |
