Assay Details
Gene Symbol: Yap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242488.2 | XP_006242550.1 | 6 - 7 | 1638 | 70 | |
XM_006242489.2 | XP_006242551.1 | 6 - 7 | 1630 | 70 | ||
XM_006242490.2 | XP_006242552.1 | 6 - 7 | 1625 | 70 | ||
XM_006242491.2 | XP_006242553.1 | 6 - 7 | 1617 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
