Assay Details
Gene Symbol: Birc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021752.2 | NP_068520.2 | 5 - 6 | 2178 | 82 | |
XM_006242494.2 | XP_006242556.1 | 6 - 7 | 2655 | 82 | ||
XM_006242495.2 | XP_006242557.1 | 6 - 7 | 2856 | 82 | ||
GenBank mRNA | AF183431.1 | - | 5 - 6 | 1536 | 82 | |
AF190020.1 | - | 5 - 6 | 2550 | 82 | ||
BC062055.1 | - | 5 - 6 | 2178 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
