Assay Details
Gene Symbol: Ecsit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242636.2 | XP_006242698.1 | 1 - 2 | 99 | 106 | |
GenBank mRNA | FQ212584.1 | - | 1 - 2 | 83 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006242636.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
