Assay Details
Gene Symbol: Ecsit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006986.1 | NP_001006987.1 | 7 - 8 | 1119 | 73 | |
XM_006242636.2 | XP_006242698.1 | 7 - 8 | 1184 | 73 | ||
XM_006242637.2 | XP_006242699.1 | 7 - 8 | 1173 | 73 | ||
XM_006242638.2 | XP_006242700.1 | 7 - 8 | 1154 | 73 | ||
GenBank mRNA | BC083762.1 | - | 7 - 8 | 1119 | 73 | |
FQ209658.1 | - | 7 - 8 | 1127 | 73 | ||
FQ215401.1 | - | 7 - 8 | 1148 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
