Assay Details
Gene Symbol: Grin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF061945.1 | - | 3 - 4 | 415 | 103 | |
AF073379.1 | - | 3402 | 103 | |||
L34938.1 | - | 2929 | 103 | |||
U29873.1 | - | 3402 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
