Assay Details
Gene Symbol: Tmeff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023020.2 | NP_075409.1 | 2 - 3 | 592 | 76 | |
GenBank mRNA | AJ250730.1 | - | 2 - 3 | 389 | 76 | |
BC129093.1 | - | 2 - 3 | 592 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
