Assay Details
Gene Symbol: Tgfbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012775.2 | NP_036907.2 | 403 | 113 | ||
GenBank mRNA | BC087035.1 | - | 345 | 113 |
Gene Symbol: LOC103690035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238153.2 | XP_006238215.2 | 1 - 2 | 1688 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
