Assay Details
Gene Symbol: Tgfbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012775.2 | NP_036907.2 | 633 | 107 | ||
XM_006238030.2 | XP_006238092.2 | 647 | 107 | |||
GenBank mRNA | BC087035.1 | - | 575 | 107 | ||
L26110.1 | - | 602 | 107 |
Gene Symbol: LOC103690035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238153.2 | XP_006238215.2 | 2 - 3 | 1918 | 107 | |
XM_008763706.1 | XP_008761928.1 | 2 - 3 | 1930 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
