Assay Details
Gene Symbol: Mxd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145773.1 | NP_665716.1 | 5 - 6 | 582 | 69 | |
GenBank mRNA | FQ234290.1 | - | 5 - 6 | 572 | 69 | |
U45986.1 | - | 5 - 6 | 582 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
