Assay Details
Gene Symbol: Sfxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012213.1 | NP_001012213.1 | 3 - 4 | 399 | 68 | |
XM_006253645.2 | XP_006253707.1 | 3 - 4 | 610 | 68 | ||
XM_008771502.1 | XP_008769724.1 | 3 - 4 | 371 | 68 | ||
GenBank mRNA | BC078935.1 | - | 3 - 4 | 399 | 68 | |
FQ209748.1 | - | 403 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
