Assay Details
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111098.1 | NP_001104568.1 | 1 - 2 | 224 | 159 | |
NM_030857.2 | NP_110484.1 | 1 - 2 | 224 | 222 | ||
GenBank mRNA | AF000300.1 | - | 1 - 2 | 69 | 222 | |
AF000301.1 | - | 1 - 2 | 224 | 159 | ||
AF000302.1 | - | 1 - 2 | 222 | 159 | ||
L14951.1 | - | 1 - 2 | 227 | 222 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF000300.1, AF000301.1, AF000302.1, L14951.1, NM_001111098.1, NM_030857.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
