Assay Details
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111098.1 | NP_001104568.1 | 2 - 3 | 294 | 140 | |
GenBank mRNA | AF000301.1 | - | 2 - 3 | 294 | 140 | |
AF000302.1 | - | 2 - 3 | 292 | 140 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF000301.1, AF000302.1, NM_001111098.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
