Assay Details
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030857.2 | NP_110484.1 | 2 - 3 | 365 | 93 | |
XM_006237834.2 | XP_006237896.1 | 2 - 3 | 681 | 93 | ||
GenBank mRNA | AF000300.1 | - | 2 - 3 | 210 | 93 | |
L14782.1 | - | 1 - 2 | 159 | 93 | ||
L14951.1 | - | 2 - 3 | 368 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
