Assay Details
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030857.2 | NP_110484.1 | 3 - 4 | 410 | 98 | |
XM_006237834.2 | XP_006237896.1 | 3 - 4 | 726 | 98 | ||
GenBank mRNA | AF000300.1 | - | 3 - 4 | 255 | 98 | |
L14782.1 | - | 2 - 3 | 204 | 98 | ||
L14951.1 | - | 3 - 4 | 413 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
